MAT047029234 Matrix Scientific

1-Allyl-1H-indole-3-carbaldehyde

  • MDL number MFCD02629520
  • Cas Number : 111480-86-5
  • Molecular Weight : 185.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 73-74
Country of Origin USA
Product Link http://www.matrixscientific.com/010380.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/010380.html
Availability details for
Quantity
  • Availability
  • Add to Cart