MAT047029257 Matrix Scientific

5-Benzylsulfanylmethyl-furan-2-carboxylic acid

  • MDL number MFCD01876278
  • Cas Number : 91903-26-3
  • Molecular Weight : 248.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 122-123
Country of Origin USA
Product Link http://www.matrixscientific.com/010401.html

Safety Information

MSDS http://www.matrixscientific.com/010401.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart