MAT047029265 Matrix Scientific

4-(1H-Indol-3-yl)-5-methyl-thiazol-2-ylamine

  • MDL number MFCD02366499
  • Cas Number : 430442-17-4
  • Molecular Weight : 229.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/010408.html

Safety Information

MSDS http://www.matrixscientific.com/010408.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart