MAT047029288 Matrix Scientific

2-Methyl-3-pyrrol-1-yl-benzoic acid

  • MDL number MFCD03080080
  • Cas Number : 83140-96-9
  • Molecular Weight : 201.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/010428.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/010428.html
Availability details for
Quantity
  • Availability
  • Add to Cart