MAT047029299 Matrix Scientific

(5-Iodo-2,4-dioxo-3,4-dihydro-2H-pyrimidin-1-yl)-acetic acid

  • MDL number MFCD01416487
  • Cas Number : 57846-83-0
  • Molecular Weight : 296.02

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 193-195
Country of Origin USA
Product Link http://www.matrixscientific.com/010438.html

Safety Information

MSDS http://www.matrixscientific.com/010438.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart