MAT047029301 Matrix Scientific

1-(2,5-Dimethoxy-phenyl)-ethylamine

  • MDL number MFCD02656663
  • Cas Number : 35253-26-0
  • Molecular Weight : 181.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/010440.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/010440.html
Availability details for
Quantity
  • Availability
  • Add to Cart