MAT047029307 Matrix Scientific

5-Cyclopropyl-7-trifluoromethyl-4,5,6,7-tetrahydro-pyrazolo[1,5-a]pyrimidine-2-carboxylic acid

  • MDL number MFCD03044654
  • Cas Number : 436088-39-0
  • Molecular Weight : 275.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/010445.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/010445.html
Availability details for
Quantity
  • Availability
  • Add to Cart