MAT047029309 Matrix Scientific

5-Isopropyl-7-trifluoromethyl-4,5,6,7-tetrahydro-pyrazolo[1,5-a]pyrimidine-2-carboxylic acid

  • MDL number MFCD03044655
  • Cas Number : 436088-40-3
  • Molecular Weight : 277.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/010447.html

Safety Information

MSDS http://www.matrixscientific.com/010447.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart