MAT047029333 Matrix Scientific

N-(4-Amino-phenyl)-3-morpholin-4-yl-propionamide

  • MDL number MFCD02737888
  • Cas Number : 462068-45-7
  • Molecular Weight : 249.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 144-146
Country of Origin USA
Product Link http://www.matrixscientific.com/010469.html

Safety Information

MSDS http://www.matrixscientific.com/010469.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart