MAT047029336 Matrix Scientific

5-{[(2-Methyl-furan-3-carbonyl)-amino]-methyl}-furan-2-carboxylic acid

  • MDL number MFCD02853136
  • Cas Number : 446054-20-2
  • Molecular Weight : 249.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 176
Country of Origin USA
Product Link http://www.matrixscientific.com/010472.html

Safety Information

MSDS http://www.matrixscientific.com/010472.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart