MAT047029345 Matrix Scientific

3-Hydroxy-2-[4-(3-methyl-butoxy)-benzoylamino]-propionic acid

  • MDL number MFCD00699953
  • Cas Number : 1008069-91-7
  • Molecular Weight : 295.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/010480.html

Safety Information

MSDS http://www.matrixscientific.com/010480.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart