MAT047029352 Matrix Scientific

10,11-Dihydro-dibenzo[b,f][1,4]oxazepine

  • MDL number MFCD00506428
  • Cas Number : 2244-60-2
  • Molecular Weight : 197.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 75-76
Country of Origin USA
Product Link http://www.matrixscientific.com/010486.html

Safety Information

MSDS http://www.matrixscientific.com/010486.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart