MAT047029364 Matrix Scientific

(4,6-Dimethyl-2,5-dioxo-hexahydro-imidazo-[4,5-d]imidazol-1-yl)-acetic acid

  • MDL number MFCD01971221
  • Cas Number : 343768-54-7
  • Molecular Weight : 228.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/010497.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/010497.html
Availability details for
Quantity
  • Availability
  • Add to Cart