MAT047029381 Matrix Scientific

4-[2-(1H-Benzoimidazol-2-yl)-ethyl]-phenylamine

  • MDL number MFCD00608303
  • Cas Number : 313524-30-0
  • Molecular Weight : 237.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 211-213
Country of Origin USA
Product Link http://www.matrixscientific.com/010513.html

Safety Information

MSDS http://www.matrixscientific.com/010513.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart