MAT047029388 Matrix Scientific

2-(1H-Benzoimidazol-5-yl)-benzooxazol-5-ylamine

  • MDL number MFCD03724111
  • Cas Number : 436086-85-0
  • Molecular Weight : 250.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/010519.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/010519.html
Availability details for
Quantity
  • Availability
  • Add to Cart