MAT047029400 Matrix Scientific

3-[5-(4-Fluoro-phenyl)-1H-pyrrol-2-yl]-propionic acid

  • MDL number MFCD03119604
  • Cas Number : 432496-81-6
  • Molecular Weight : 233.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/010528.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/010528.html
Availability details for
Quantity
  • Availability
  • Add to Cart