MAT047029403 Matrix Scientific

3-(5-p-Tolyl-furan-2-yl)-propionic acid

  • MDL number MFCD03110272
  • Cas Number : 23589-06-2
  • Molecular Weight : 230.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/010531.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/010531.html
Availability details for
Quantity
  • Availability
  • Add to Cart