MAT047029410 Matrix Scientific

7-(4-Methoxy-phenyl)-4,7-dioxo-heptanoic acid

  • MDL number MFCD02257796
  • Cas Number : 24090-40-2
  • Molecular Weight : 264.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 119
Country of Origin USA
Product Link http://www.matrixscientific.com/010537.html

Safety Information

MSDS http://www.matrixscientific.com/010537.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart