MAT047029415 Matrix Scientific

Benzofuran-2-carboxylic acid (4-amino-2-methyl-phenyl)-amide

  • MDL number MFCD03030599
  • Cas Number : 433252-24-5
  • Molecular Weight : 266.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/010542.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/010542.html
Availability details for
Quantity
  • Availability
  • Add to Cart