MAT047029417 Matrix Scientific

5-Chloro-2-(trifluoromethyl)phenylacetic acid

  • MDL number MFCD01631358
  • Cas Number : 261763-25-1
  • Molecular Weight : 238.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 117-119
Country of Origin USA
Product Link http://www.matrixscientific.com/002006.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/002006.html
Availability details for
Quantity
  • Availability
  • Add to Cart