MAT047029418 Matrix Scientific

4-(Furan-2-ylmethylsulfanylmethyl)-benzoic acid

  • MDL number MFCD01418359
  • Cas Number : 312517-86-5
  • Molecular Weight : 248.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/010544.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/010544.html
Availability details for
Quantity
  • Availability
  • Add to Cart