MAT047029420 Matrix Scientific

(4-Methoxy-phenyl)-pyridin-4-ylmethyl-amine

  • MDL number MFCD00592910
  • Cas Number : 16552-49-1
  • Molecular Weight : 214.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 98
Country of Origin USA
Product Link http://www.matrixscientific.com/010546.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/010546.html
Availability details for
Quantity
  • Availability
  • Add to Cart