MAT047029428 Matrix Scientific

5-Chloro-2-(trifluoromethyl)phenylacetonitrile

  • MDL number MFCD01631357
  • Cas Number : 261763-26-2
  • Molecular Weight : 219.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/002007.html

Safety Information

hcodes Toxic
MSDS http://www.matrixscientific.com/002007.html
Availability details for
Quantity
  • Availability
  • Add to Cart