MAT047029438 Matrix Scientific

[2-(4-Methoxy-phenyl)-ethyl]-(4-methylsulfanyl-benzyl)-amine

  • MDL number MFCD01135448
  • Cas Number : 331970-71-9
  • Molecular Weight : 287.43

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/010563.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/010563.html
Availability details for
Quantity
  • Availability
  • Add to Cart