MAT047029439 Matrix Scientific

3-Fluoro-2-(trifluoromethyl)benzamide

  • MDL number MFCD01631492
  • Cas Number : 287398-80-5
  • Molecular Weight : 207.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://www.matrixscientific.com/002008.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/002008.html
Availability details for
Quantity
  • Availability
  • Add to Cart