MAT047029461 Matrix Scientific

3-Fluoro-2-(trifluoromethyl)benzonitrile

  • MDL number MFCD01631566
  • Cas Number : 261951-81-9
  • Molecular Weight : 189.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/002010.html

Safety Information

hcodes Toxic/Irritant
MSDS http://www.matrixscientific.com/002010.html
Availability details for
Quantity
  • Availability
  • Add to Cart