MAT047029468 Matrix Scientific

Furan-2-ylmethyl-[1-(4-methoxy-phenyl)-3-methyl-but-3-enyl]-amine

  • MDL number MFCD03274721
  • Cas Number : 436088-84-5
  • Molecular Weight : 271.36

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/010590.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/010590.html
Availability details for
Quantity
  • Availability
  • Add to Cart