MAT047029480 Matrix Scientific

(2-Chloro-benzyl)-pyridin-3-ylmethyl-amine

  • MDL number MFCD02609563
  • Cas Number : 436088-71-0
  • Molecular Weight : 232.71

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/010601.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/010601.html
Availability details for
Quantity
  • Availability
  • Add to Cart