MAT047029489 Matrix Scientific

3-(3-Methyl-pyrazol-1-yl)-propylamine

  • MDL number MFCD03194398
  • Cas Number : 62821-91-4
  • Molecular Weight : 139.20

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Boiling point 106-109� Torr
Product Link http://www.matrixscientific.com/010609.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/010609.html
Availability details for
Quantity
  • Availability
  • Add to Cart