MAT047029499 Matrix Scientific

Benzo[1,3]dioxol-5-ylmethyl-benzyl-amine

  • MDL number MFCD00444190
  • Cas Number : 4720-73-4
  • Molecular Weight : 241.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Boiling point 130�25 Torr
Product Link http://www.matrixscientific.com/010618.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/010618.html
Availability details for
Quantity
  • Availability
  • Add to Cart