MAT047029501 Matrix Scientific

Benzyl-(4-fluoro-benzyl)-amine

  • MDL number MFCD00716882
  • Cas Number : 55096-88-3
  • Molecular Weight : 215.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 62
Country of Origin USA
Product Link http://www.matrixscientific.com/010620.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/010620.html
Availability details for
Quantity
  • Availability
  • Add to Cart