MAT047029505 Matrix Scientific

3,5-Bis(1,1,1,3,3,3-hexafluoro-2-hydroxypropyl)-iodobenzene

  • MDL number MFCD04038221
  • Cas Number : 53173-72-1
  • Molecular Weight : 536.06

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Boiling point 115� Torr
Product Link http://www.matrixscientific.com/001121.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/001121.html
Availability details for
Quantity
  • Availability
  • Add to Cart