MAT047029509 Matrix Scientific

Benzyl-(2,5-dimethoxy-benzyl)-amine

  • MDL number MFCD03821856
  • Cas Number : 626205-84-3
  • Molecular Weight : 257.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/010626.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/010626.html
Availability details for
Quantity
  • Availability
  • Add to Cart