MAT047029510 Matrix Scientific

Benzyl-(2,3,4-trimethoxy-benzyl)-amine

  • MDL number MFCD04035006
  • Cas Number : 436086-80-5
  • Molecular Weight : 287.36

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/010627.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/010627.html
Availability details for
Quantity
  • Availability
  • Add to Cart