MAT047029518 Matrix Scientific

(4-Fluorobenzyl)-pyridin-3-ylmethyl-amine

  • MDL number MFCD02656616
  • Cas Number : 113248-64-9
  • Molecular Weight : 216.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/010634.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/010634.html
Availability details for
Quantity
  • Availability
  • Add to Cart