MAT047029523 Matrix Scientific

(2-Fluoro-benzyl)-pyridin-3-ylmethyl-amine

  • MDL number MFCD03176052
  • Cas Number : 500221-74-9
  • Molecular Weight : 216.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/010639.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/010639.html
Availability details for
Quantity
  • Availability
  • Add to Cart