MAT047029525 Matrix Scientific

(2,3-Dimethoxy-benzyl)-pyridin-3-ylmethyl-amine

  • MDL number MFCD01135933
  • Cas Number : 355383-37-8
  • Molecular Weight : 258.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/010641.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/010641.html
Availability details for
Quantity
  • Availability
  • Add to Cart