MAT047029535 Matrix Scientific

(4-Bromo-benzyl)-pyridin-3-ylmethyl-amine

  • MDL number MFCD03724418
  • Cas Number : 510723-60-1
  • Molecular Weight : 277.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/010650.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/010650.html
Availability details for
Quantity
  • Availability
  • Add to Cart