MAT047029536 Matrix Scientific

Pyridin-3-ylmethyl-(tetrahydro-furan-2-ylmethyl)-amine

  • MDL number MFCD03001390
  • Cas Number : 519144-57-1
  • Molecular Weight : 192.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/010651.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/010651.html
Availability details for
Quantity
  • Availability
  • Add to Cart