MAT047029551 Matrix Scientific

Benzo[1,3]dioxol-5-ylmethyl-furan-2-ylmethyl-amine

  • MDL number MFCD00444189
  • Cas Number : 510723-65-6
  • Molecular Weight : 231.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/010664.html

Safety Information

MSDS http://www.matrixscientific.com/010664.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart