MAT047029563 Matrix Scientific

Furan-2-ylmethyl-(3-methoxy-benzyl)-amine

  • MDL number MFCD03724429
  • Cas Number : 510723-73-6
  • Molecular Weight : 217.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/010675.html

Safety Information

MSDS http://www.matrixscientific.com/010675.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart