MAT047029569 Matrix Scientific

(2-Chloro-benzyl)-(tetrahydro-furan-2-ylmethyl)-amine

  • MDL number MFCD03210836
  • Cas Number : 356532-08-6
  • Molecular Weight : 225.72

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/010681.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/010681.html
Availability details for
Quantity
  • Availability
  • Add to Cart