MAT047029573 Matrix Scientific

(4-Isopropyl-benzyl)-(tetrahydro-furan-2-yl-methyl)-amine

  • MDL number MFCD03446308
  • Cas Number : 436096-82-1
  • Molecular Weight : 233.36

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/010684.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/010684.html
Availability details for
Quantity
  • Availability
  • Add to Cart