MAT047029575 Matrix Scientific

(4-Fluoro-benzyl)-(tetrahydro-furan-2-ylmethyl)-amine

  • MDL number MFCD02656620
  • Cas Number : 356531-65-2
  • Molecular Weight : 209.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/010686.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/010686.html
Availability details for
Quantity
  • Availability
  • Add to Cart