MAT047029578 Matrix Scientific

(2-Chloro-6-fluoro-benzyl)-(tetrahydro-furan-2-yl-methyl)-amine

  • MDL number MFCD03724432
  • Cas Number : 510723-77-0
  • Molecular Weight : 243.71

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/010689.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/010689.html
Availability details for
Quantity
  • Availability
  • Add to Cart