MAT047029604 Matrix Scientific

3-(Quinoline-8-sulfonyl)-propionic acid

  • MDL number MFCD03448765
  • Cas Number : 626206-92-6
  • Molecular Weight : 265.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 198-200
Country of Origin USA
Product Link http://www.matrixscientific.com/010713.html

Safety Information

MSDS http://www.matrixscientific.com/010713.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart