MAT047029606 Matrix Scientific

1-(4-Methoxy-phenyl)-2-p-tolylamino-ethanol

  • MDL number MFCD03724597
  • Cas Number : 510737-64-1
  • Molecular Weight : 257.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/010714.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/010714.html
Availability details for
Quantity
  • Availability
  • Add to Cart