MAT047029610 Matrix Scientific

3-(2-Morpholin-4-yl-acetylamino)-benzoic acid

  • MDL number MFCD01170920
  • Cas Number : 446852-20-6
  • Molecular Weight : 264.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/010718.html

Safety Information

MSDS http://www.matrixscientific.com/010718.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart