MAT047029618 Matrix Scientific

4-Aminomethyl-5-methyl-furan-2-carboxylic acid

  • MDL number MFCD03274774
  • Cas Number : 436088-95-8
  • Molecular Weight : 155.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/010724.html

Safety Information

MSDS http://www.matrixscientific.com/010724.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart