MAT047029636 Matrix Scientific

3-(2-Fluoro-benzylamino)-propan-1-ol

  • MDL number MFCD03724652
  • Cas Number : 436087-21-7
  • Molecular Weight : 183.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/010741.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/010741.html
Availability details for
Quantity
  • Availability
  • Add to Cart